Learn More
Dimedone, 98%, Thermo Scientific Chemicals
Description
Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.
Solubility
Soluble in methanol, ethanol, benzene and chloroform. Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents, strong acids and strong bases.
Specifications
Melting Point | 146°C to 150°C |
Quantity | 25 g |
Beilstein | 471489 |
Merck Index | 14,3242 |
Solubility Information | Soluble in methanol,ethanol,benzene and chloroform. Slightly soluble in water. |
Formula Weight | 140.18 |
Percent Purity | 98% |
Chemical Name or Material | Dimedone |
Safety and Handling
EINECSNumber | 204-804-4 |
RTECSNumber | GV0345000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.