Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimenhydrinate, USP, 53-55.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
523-87-5
C24H28ClN5O3
469.97
NFLLKCVHYJRNRH-UHFFFAOYSA-N
8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; [2-(diphenylmethoxy)ethyl]dimethylamine
CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O.CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
523-87-5
C24H28ClN5O3
469.97
NFLLKCVHYJRNRH-UHFFFAOYSA-N
8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; [2-(diphenylmethoxy)ethyl]dimethylamine
CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O.CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

DI127, 523-87-5, C17H21NO•C7H7ClN4O2

Spectrum™ Dimenhydrinate, USP is used to prevent and treat nausea, vomiting, and dizziness caused by motion sickness. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Quantity 1 kg
Ignition Residue 0.003
CAS Max % 1
Percent Purity 53 to 55.5%