Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Dimenthyl fumarate, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
34675-24-6
C24H40O4
392.58
HTJZKHLYRXPLLS-RKGYUCNASA-N
--di 1r-menthyl fumarate,di-1r-menthyl fumarate
18397611
bis[(5R)-5-methyl-2-propan-2-ylcyclohexyl] (E)-but-2-enedioate
CC1CCC(C(C1)OC(=O)C=CC(=O)OC2CC(CCC2C(C)C)C)C(C)C
34675-24-6
C24H40O4
392.58
HTJZKHLYRXPLLS-RKGYUCNASA-N
--di 1r-menthyl fumarate,di-1r-menthyl fumarate
18397611
bis[(5R)-5-methyl-2-propan-2-ylcyclohexyl] (E)-but-2-enedioate
CC1CCC(C(C1)OC(=O)C=CC(=O)OC2CC(CCC2C(C)C)C)C(C)C
Boiling Point 211°C (7.0 hPa)
Specific Rotation − 103.14
Quantity 5g
Infrared Spectrum Authentic
Melting Point 59.0°C to 61.0°C
Color Colorless to White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,acetone,hexane
Formula Weight 392.58
Physical Form Low Melting Solid or Powder
Percent Purity 95%
Chemical Name or Material (-)-Dimenthyl fumarate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning