Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxydimethylsilane, 95+%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
1112-39-6
C4H12O2Si
120.23
JJQZDUKDJDQPMQ-UHFFFAOYSA-N
dimethyldimethoxysilane,silane, dimethoxydimethyl,kbm 22,unii-a3qdb1rs1c,dimethyl dimethoxysilane,dimethoxy dimethyl silane,a3qdb1rs1c,dimethoxy-dimethylsilane,dimethyl dimethoxy silane,dimethyl dimethoxy silicane
66187
dimethoxy(dimethyl)silane
CO[Si](C)(C)OC
1112-39-6
C4H12O2Si
120.23
JJQZDUKDJDQPMQ-UHFFFAOYSA-N
dimethyldimethoxysilane,silane, dimethoxydimethyl,kbm 22,unii-a3qdb1rs1c,dimethyl dimethoxysilane,dimethoxy dimethyl silane,a3qdb1rs1c,dimethoxy-dimethylsilane,dimethyl dimethoxy silane,dimethyl dimethoxy silicane
66187
dimethoxy(dimethyl)silane
CO[Si](C)(C)OC
Refractive Index 1.3690 to 1.3710
Specific Gravity 0.865
Packaging AcroSeal™ Glass Bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Melting Point -80.0°C
Flash Point −8°C
Boiling Point 81.0°C to 82.0°C
Quantity 1L
Density 0.8650g/mL
Assay Percent Range 95% min. (GC)
Linear Formula (CH3)2Si(OCH3)2
Solubility Information Solubility in water: reacts with water to form methanol
Formula Weight 120.23
Physical Form Liquid
Percent Purity 95+%
Chemical Name or Material Dimethoxydimethylsilane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Danger

EINECSNumber : 214-189-4