Learn More
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals
Description
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.
Solubility
Soluble in water 0.0016 g/L.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | 152°C to 156°C |
Quantity | 500 g |
Solubility Information | Soluble in water 0.0016g/L. |
IUPAC Name | 1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate |
Formula Weight | 228.2 |
Percent Purity | ≥99% |
Chemical Name or Material | Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 228-528-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.