Alfa Aesar™ Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%

Manufacturer: Alfa Aesar™ B2400822

Quantity 100g

 View more versions of this product

Catalog No. AAB2400822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6289-46-9
    Molecular Formula C10H12O6
    Formula Weight 228.2
    MDL Number MFCD00001607
    Synonym 1,4-Cyclohexanedione-2,5-dicarboxylic acid dimethyl ester; Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
    Chemical Name or Material Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate

    Specifications

    Formula Weight 228.2
    Melting Point 152° to 156°C
    TSCA TSCA
    Molecular Formula C10H12O6
    MDL Number MFCD00001607
    CAS 6289-46-9
    Quantity 100g
    Synonym 1,4-Cyclohexanedione-2,5-dicarboxylic acid dimethyl ester; Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
    Chemical Name or Material Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
    Assay ≥99%

    Description

    Applications
    Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

    Solubility
    Soluble in water 0.0016 g/L.

    Notes
    Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit