Alfa Aesar™ Dimethyl (1-ethylpropyl)malonate, 97%

Manufacturer: Alfa Aesar™ H26701ME

Quantity 500mg

 View more versions of this product

Catalog No. AAH26701ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39520-19-9
    Molecular Formula C10H18O4
    Formula Weight 202.25
    MDL Number MFCD08436116
    Synonym Dimethyl (diethylmethyl)malonate; (1-Ethylpropyl)malonic acid dimethyl ester
    Chemical Name or Material Dimethyl (1-ethylpropyl)malonate

    Specifications

    Formula Weight 202.25
    Molecular Formula C10H18O4
    MDL Number MFCD08436116
    CAS 39520-19-9
    Quantity 500mg
    Synonym Dimethyl (diethylmethyl)malonate; (1-Ethylpropyl)malonic acid dimethyl ester
    Chemical Name or Material Dimethyl (1-ethylpropyl)malonate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit