Alfa Aesar™ (-)-Dimethyl 2,3-O-isopropylidene-L-tartrate, 94%

Manufacturer: Alfa Aesar™ H27220AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH27220AA

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 37031-29-1
    Molecular Formula C9H14O6
    Formula Weight 218.2
    MDL Number MFCD00066218
    Synonym Dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
    Chemical Name or Material (-)-Dimethyl 2,3-O-isopropylidene-L-tartrate

    Specifications

    Formula Weight 218.2
    Molecular Formula C9H14O6
    Boiling Point 80° to 82°C (0.1mmHg)
    MDL Number MFCD00066218
    CAS 37031-29-1
    Quantity 5mL
    Flash Point >110°C (230°F)
    Synonym Dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
    Chemical Name or Material (-)-Dimethyl 2,3-O-isopropylidene-L-tartrate
    Assay 94%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit