Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl 2-fluoroterephthalate, 98%

Manufacturer:  Alfa Aesar™ 4384906

Quantity 5g

 View more versions of this product

Catalog No. AA4384906


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

5292-47-7
212.17
Dimethyl 2-fluoroterephthalate
C10H9FO4
MFCD00042266

Specifications

5292-47-7
C10H9FO4
212.17
5g
98%
Powder
81° to 83°C
MFCD00042266
Dimethyl 2-fluoroterephthalate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit