Alfa Aesar™ Dimethyl 2-fluoroterephthalate, 98%

Manufacturer: Alfa Aesar™ 4384903

Quantity 1g

 View more versions of this product

Catalog No. AA4384903

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 5292-47-7
    Molecular Formula C10H9FO4
    Formula Weight 212.17
    MDL Number MFCD00042266
    Chemical Name or Material Dimethyl 2-fluoroterephthalate

    Specifications

    Formula Weight 212.17
    Molecular Formula C10H9FO4
    Physical Form Powder
    Melting Point 81° to 83°C
    MDL Number MFCD00042266
    Quantity 1g
    CAS 5292-47-7
    Chemical Name or Material Dimethyl 2-fluoroterephthalate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit