Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 2-hydroxyethylphosphonate, 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
54731-72-5
C4H11O4P
154.1
MFCD00043376
TZPPDWDHNIMTDQ-UHFFFAOYSA-N
dimethyl 2-hydroxyethylphosphonate,dimethyl 2-hydroxyethyl phosphonate,2-hydroxyethylphosphonic acid dimethyl ester,2-hydroxyethyl phosphonic acid dimethyl ester,acmc-1ao1t,2-diethylphosphono ethan-1-ol,tzppdwdhnimtdq-uhfffaoysa,hydroxyethyldimethylphosphonic acid,2-dimethoxyphosphoryl ethan-1-ol
2733833
2-dimethoxyphosphorylethanol
COP(=O)(CCO)OC
54731-72-5
C4H11O4P
154.1
MFCD00043376
TZPPDWDHNIMTDQ-UHFFFAOYSA-N
dimethyl 2-hydroxyethylphosphonate,dimethyl 2-hydroxyethyl phosphonate,2-hydroxyethylphosphonic acid dimethyl ester,2-hydroxyethyl phosphonic acid dimethyl ester,acmc-1ao1t,2-diethylphosphono ethan-1-ol,tzppdwdhnimtdq-uhfffaoysa,hydroxyethyldimethylphosphonic acid,2-dimethoxyphosphoryl ethan-1-ol
2733833
2-dimethoxyphosphorylethanol
COP(=O)(CCO)OC
for C-C Bond Formation (Olefination)
Refractive Index 1.4430 to 1.4460
Specific Gravity 1.247
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 25mL
Flash Point 192°C
Density 1.2470g/mL
Boiling Point 98.0°C to 104.0°C (0.1mmHg)
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Linear Formula (CH3O)2P(O)(CH2)2OH
Beilstein 04, IV, 3584
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 154.1
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Dimethyl 2-hydroxyethylphosphonate, 95%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-310-1