Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl 4-chloropyridine-2,6-dicarboxylate, 98%

Click to view available options
Quantity:
250mg
1g
5g
5371-70-0
C9H8ClNO4
229.616
MFCD02612781
NFXKYKHKNUFOKB-UHFFFAOYSA-N
dimethyl 4-chloro-2,6-pyridinedicarboxylate,methyl 4-chloropyridine-2,6-dicarboxylate,2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate,2,6-pyridinedicarboxylic acid, 4-chloro-, dimethyl ester,4-chloropyridine-2,6-dicarboxylic acid,dimethyl ester,methyl 4-chloro-6-methoxycarbonyl pyridine-2-carboxylate,4-chloro-2,6-pyridinedicarboxylic acid dimethyl ester,pubchem20951,pych37,ksc496e3d
237845
dimethyl 4-chloropyridine-2,6-dicarboxylate
COC(=O)C1=CC(=CC(=N1)C(=O)OC)Cl
5371-70-0
C9H8ClNO4
229.616
MFCD02612781
NFXKYKHKNUFOKB-UHFFFAOYSA-N
dimethyl 4-chloro-2,6-pyridinedicarboxylate,methyl 4-chloropyridine-2,6-dicarboxylate,2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate,2,6-pyridinedicarboxylic acid, 4-chloro-, dimethyl ester,4-chloropyridine-2,6-dicarboxylic acid,dimethyl ester,methyl 4-chloro-6-methoxycarbonyl pyridine-2-carboxylate,4-chloro-2,6-pyridinedicarboxylic acid dimethyl ester,pubchem20951,pych37,ksc496e3d
237845
dimethyl 4-chloropyridine-2,6-dicarboxylate
COC(=O)C1=CC(=CC(=N1)C(=O)OC)Cl
Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color Yellow
Melting Point 139°C to 144°C
Quantity 1g
Beilstein 197075
Formula Weight 229.62
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Dimethyl 4-chloropyridine-2,6-dicarboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning