Alfa Aesar™ Dimethyl 5-nitroisophthalate, 98+%

Manufacturer: Alfa Aesar™ A1061018

Quantity 50g

 View more versions of this product

Catalog No. AAA1061018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13290-96-5
    Molecular Formula C10H9NO6
    Formula Weight 239.19
    MDL Number MFCD00008429
    Beilstein 2140916
    Synonym 5-Nitrobenzene-1,3-dicarboxylic acid dimethyl ester; 5-Nitroisophthalic acid dimethyl ester
    Chemical Name or Material Dimethyl 5-nitroisophthalate

    Specifications

    Formula Weight 239.19
    CAS 13290-96-5
    Molecular Formula C10H9NO6
    Melting Point 121° to 123°C
    Beilstein 2140916
    Synonym 5-Nitrobenzene-1,3-dicarboxylic acid dimethyl ester; 5-Nitroisophthalic acid dimethyl ester
    RTECS Number CZ4340000
    TSCA TSCA
    MDL Number MFCD00008429
    Quantity 50g
    Chemical Name or Material Dimethyl 5-nitroisophthalate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit