Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl 5-sulfoisophthalate sodium salt, 98%

Click to view available options
Quantity:
25g
100g
500g
3965-55-7
C10H9NaO7S
296.225
MFCD00007493
LLHSEQCZSNZLRI-UHFFFAOYSA-M
sodium 3,5-bis methoxycarbonyl benzenesulfonate,sodium dimethyl 5-sulphonatoisophthalate,dimethyl 5-sulfoisophthalate sodium salt,unii-7l3g5x9vdu,sodium 1,3-dimethyl 5-sulfoisophthalate,7l3g5x9vdu,5-sulfoisophthalic acid, dimethyl ester, sodium salt,sodium dimethyl 5-sulfoisophthalate,1,3-benzenedicarboxylic acid, 5-sulfo-, 1,3-dimethyl ester, sodium salt,3,5-bis-methylkarboxy-benzensulfonan sodny czech
23675796
sodium;3,5-bis(methoxycarbonyl)benzenesulfonate
COC(=O)C1=CC(=CC(=C1)S(=O)(=O)[O-])C(=O)OC.[Na+]
3965-55-7
C10H9NaO7S
296.225
MFCD00007493
LLHSEQCZSNZLRI-UHFFFAOYSA-M
sodium 3,5-bis methoxycarbonyl benzenesulfonate,sodium dimethyl 5-sulphonatoisophthalate,dimethyl 5-sulfoisophthalate sodium salt,unii-7l3g5x9vdu,sodium 1,3-dimethyl 5-sulfoisophthalate,7l3g5x9vdu,5-sulfoisophthalic acid, dimethyl ester, sodium salt,sodium dimethyl 5-sulfoisophthalate,1,3-benzenedicarboxylic acid, 5-sulfo-, 1,3-dimethyl ester, sodium salt,3,5-bis-methylkarboxy-benzensulfonan sodny czech
23675796
sodium;3,5-bis(methoxycarbonyl)benzenesulfonate
COC(=O)C1=CC(=CC(=C1)S(=O)(=O)[O-])C(=O)OC.[Na+]
Melting Point >300°C
Quantity 25g
Formula Weight 296.23
Percent Purity 98%
Chemical Name or Material Dimethyl 5-sulfoisophthalate sodium salt

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA