Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl adipate, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
627-93-0
C8H14O4
174.2
MFCD00008469
UDSFAEKRVUSQDD-UHFFFAOYSA-N
dimethyl adipate,dimethyladipate,adipic acid dimethyl ester,hexanedioic acid, dimethyl ester,adipic acid, dimethyl ester,1,6-dimethylhexanedioate,methyl adipate van,1,6-dimethyl hexanedioate,dbe 6,unii-by71rx0r62
12329
CHEBI:34715
dimethyl hexanedioate
COC(=O)CCCCC(=O)OC
627-93-0
C8H14O4
174.2
MFCD00008469
UDSFAEKRVUSQDD-UHFFFAOYSA-N
dimethyl adipate,dimethyladipate,adipic acid dimethyl ester,hexanedioic acid, dimethyl ester,adipic acid, dimethyl ester,1,6-dimethylhexanedioate,methyl adipate van,1,6-dimethyl hexanedioate,dbe 6,unii-by71rx0r62
12329
CHEBI:34715
dimethyl hexanedioate
COC(=O)CCCCC(=O)OC
Viscosity 5 mPa.s (20°C)
Density 1.0600g/mL
Specific Gravity 1.06
Water 0.1% max. (K.F.)
Packaging Glass bottle
Refractive Index 1.4270 to 1.4290
Color Undesignated
Infrared Spectrum Authentic
Boiling Point 109.0°C to 110.0°C (14.0 mmHg)
Quantity 100g
Flash Point 107°C
Acidity 0.01% max. (as adipic acid)
Melting Point 8.0°C
Assay Percent Range 99% min. (GC)
Linear Formula CH3O2C(CH2)4CO2CH3
Beilstein 02, 652
Merck Index 10, 151
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,alcohols,carbon tetrachloride,,acetic acid
Formula Weight 174.2
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Dimethyl adipate, 99+%
Show More Show Less

EINECSNumber : 211-020-6

RTECSNumber : AV1645000

TSCA : TSCA