Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl aminoterephthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
50g
250g
Packaging:
Plastic bottle
5372-81-6
C10H11NO4
209.2
MFCD00008427
DSSKDXUDARIMTR-UHFFFAOYSA-N
dimethyl 2-aminoterephthalate,dimethyl aminoterephthalate,dimethyl 3-aminoterephthalate,1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester,aminoterephthalic acid dimethyl ester,1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate,unii-91sf4e6i9w,2-aminoterephthalic acid, dimethyl ester,3-amino-4-methoxycarbonylbenzoic acid, methyl ester,terephthalic acid, amino-, dimethyl ester
79336
dimethyl 2-aminobenzene-1,4-dicarboxylate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)N
5372-81-6
C10H11NO4
209.2
MFCD00008427
DSSKDXUDARIMTR-UHFFFAOYSA-N
dimethyl 2-aminoterephthalate,dimethyl aminoterephthalate,dimethyl 3-aminoterephthalate,1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester,aminoterephthalic acid dimethyl ester,1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate,unii-91sf4e6i9w,2-aminoterephthalic acid, dimethyl ester,3-amino-4-methoxycarbonylbenzoic acid, methyl ester,terephthalic acid, amino-, dimethyl ester
79336
dimethyl 2-aminobenzene-1,4-dicarboxylate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)N
Packaging Plastic bottle
Infrared Spectrum Authentic
Melting Point 132.0°C to 134.0°C
Quantity 250g
Color Beige or Brown to Yellow
Assay Percent Range 98.5% min. (GC)
Linear Formula H2NC6H3(CO2CH3)2
Beilstein 14, 559
Formula Weight 209.2
Physical Form Powder or Crystals
Percent Purity 99%
Chemical Name or Material Dimethyl aminoterephthalate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-364-2

TSCA : TSCA