Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Carbonate, 99%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Steel Can
616-38-6
616-38-6
Packaging Glass Bottle
Quantity 2.5 kg
CAS Max % 1
Percent Purity 0.99

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.