Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Chlorothiophosphate 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
2524-03-0
C2H6ClO2PS
160.552
MFCD00008353
XFBJRFNXPUCPKU-UHFFFAOYSA-N
dimethyl chlorothiophosphate,methyl pct,o,o-dimethyl chlorothiophosphate,o,o-dimethyl phosphorochloridothioate,phosphorochloridothioic acid, o,o-dimethyl ester,chlorodimethoxyphosphine sulfide,dimethylchlorthiofosfat,dimethyl chlorothionophosphate,dimethyl chlorothiophosphonate,dimethyl phosphorochlorothioate
17304
chloro-dimethoxy-sulfanylidene-$l^{5}-phosphane
COP(=S)(OC)Cl
2524-03-0
C2H6ClO2PS
160.552
MFCD00008353
XFBJRFNXPUCPKU-UHFFFAOYSA-N
dimethyl chlorothiophosphate,methyl pct,o,o-dimethyl chlorothiophosphate,o,o-dimethyl phosphorochloridothioate,phosphorochloridothioic acid, o,o-dimethyl ester,chlorodimethoxyphosphine sulfide,dimethylchlorthiofosfat,dimethyl chlorothionophosphate,dimethyl chlorothiophosphonate,dimethyl phosphorochlorothioate
17304
chloro-dimethoxy-sulfanylidene-$l^{5}-phosphane
COP(=S)(OC)Cl
Packaging Glass bottle
Refractive Index 1.4810 to 1.4830 (20°C, 589nm)
Quantity 100g
Color Colorless to Yellow
Infrared Spectrum Authentic
Density 1.32g/mL
Assay Percent Range 97%
Linear Formula (CH3O)2P(S)Cl
Beilstein 01, I, 143
Formula Weight 160.55
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Dimethyl chlorothiophosphate, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Fatal if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-754-9

RTECSNumber : TD1830000

TSCA : TSCA