Alfa Aesar™ Dimethyl cis-4-cyclohexene-1,2-dicarboxylate, 99%

Manufacturer: Alfa Aesar™ H2595203

Quantity 1g

 View more versions of this product

Catalog No. AAH2595203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4841-84-3
    Molecular Formula C10H14O4
    Formula Weight 198.22
    MDL Number MFCD00209581
    Synonym cis-4-Cyclohexene-1,2-dicarboxylic acid dimethyl ester; Dimethyl cis-1,2,3,6-tetrahydrophthalate
    Chemical Name or Material Dimethyl cis-4-cyclohexene-1,2-dicarboxylate

    Specifications

    Formula Weight 198.22
    Molecular Formula C10H14O4
    Boiling Point 141° to 142°C (20mmHg)
    MDL Number MFCD00209581
    CAS 4841-84-3
    Quantity 1g
    Flash Point >110°C (230°F)
    Synonym cis-4-Cyclohexene-1,2-dicarboxylic acid dimethyl ester; Dimethyl cis-1,2,3,6-tetrahydrophthalate
    Chemical Name or Material Dimethyl cis-4-cyclohexene-1,2-dicarboxylate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit