Alfa Aesar™ Dimethyl diselenide, 99%

Manufacturer: Alfa Aesar™ 6811903

Quantity 1g

 View more versions of this product

Catalog No. AA6811903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7101-31-7
    Molecular Formula (CH3)2Se2
    Formula Weight 187.99
    MDL Number MFCD00008254
    UN Number UN2929
    Synonym Methyl diselenide
    Chemical Name or Material Dimethyl diselenide

    Specifications

    Formula Weight 187.99
    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    CAS 7101-31-7
    Synonym Methyl diselenide
    Quantity 1g
    Density 1.987g/mL
    Sensitivity Air & Moisture Sensitive
    Odor Unpleasant
    Molecular Formula (CH3)2Se2
    Refractive Index 1.6388
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
    Flash Point 60°C (140°F)
    Boiling Point 155° to 157°C
    MDL Number MFCD00008254
    UN Number UN2929
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331-H226-H373
    Toxic if swallowed.
    Toxic if inhaled.
    Flammable liquid and vapor.
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Dimethyl diselenide
    Assay 99%

    GHS H Statement
    H301-H331-H226-H373
    Toxic if swallowed.
    Toxic if inhaled.
    Flammable liquid and vapor.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit