Alfa Aesar™ Dimethyl glutarate, 98%

Manufacturer: Alfa Aesar™ B24359-36

Quantity 500g

 View more versions of this product

Catalog No. AAB2435936

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1119-40-0
    Molecular Formula C7H12O4
    Formula Weight 160.17
    MDL Number MFCD00008468
    Beilstein 1771444
    Merck Index 14,4473
    Synonym Glutaric acid dimethyl ester; Pentanedioic acid dimethyl ester
    Chemical Name or Material Dimethyl glutarate

    Specifications

    Formula Weight 160.17
    MDL Number MFCD00008468
    CAS 1119-40-0
    Boiling Point 213° to 215°C
    Quantity 500g
    TSCA TSCA
    Molecular Formula C7H12O4
    Beilstein 1771444
    Merck Index 14,4473
    Synonym Glutaric acid dimethyl ester; Pentanedioic acid dimethyl ester
    Flash Point 103°C (217°F)
    Melting Point −37°C
    Chemical Name or Material Dimethyl glutarate
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit