Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Glutarate 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
1119-40-0
C7H12O4
160.17
MFCD00008468
XTDYIOOONNVFMA-UHFFFAOYSA-N
dimethyl glutarate,glutaric acid dimethyl ester,pentanedioic acid, dimethyl ester,methyl glutarate,glutaric acid, dimethyl ester,dbe 5,unii-1i9vfa346p,pentanedioic acid, 1,5-dimethyl ester,1,5-dimethyl pentanedioate,pentanedioic acid dimethyl ester
14242
dimethyl pentanedioate
COC(=O)CCCC(=O)OC
1119-40-0
C7H12O4
160.17
MFCD00008468
XTDYIOOONNVFMA-UHFFFAOYSA-N
dimethyl glutarate,glutaric acid dimethyl ester,pentanedioic acid, dimethyl ester,methyl glutarate,glutaric acid, dimethyl ester,dbe 5,unii-1i9vfa346p,pentanedioic acid, 1,5-dimethyl ester,1,5-dimethyl pentanedioate,pentanedioic acid dimethyl ester
14242
dimethyl pentanedioate
COC(=O)CCCC(=O)OC
Viscosity 2.5 mm2/s (25°C)
Density 1.0870g/mL
Melting Point -37.0°C
Specific Gravity 1.087
Packaging Glass bottle
Refractive Index 1.4236 to 1.4256
Infrared Spectrum Authentic
Quantity 500g
Flash Point 103°C
Boiling Point 210.0°C to 215.0°C
Color Colorless
Assay Percent Range 98%
Linear Formula CH3O2C(CH2)3CO2CH3
Beilstein 02, 633
Merck Index 15, 4509
Solubility Information Solubility in water: 53g/L
Formula Weight 160.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Dimethyl glutarate, 98%
Show More Show Less

EINECSNumber : 214-277-2

TSCA : TSCA