Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl isobutylmalonate, 98%, Thermo Scientific™

Catalog Number ac350470050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
39520-24-6
C9H16O4
188.22
MFCD00015640
GAZFDPSEEIVCEX-UHFFFAOYSA-N
dimethyl isobutylmalonate, dimethyl 2-isobutylmalonate, dimethylisobutylmalonate, propanedioic acid, 2-2-methylpropyl-, 1,3-dimethyl ester, propanedioic acid, 2-methylpropyl-, dimethyl ester, 1,3-dimethyl 2-2-methylpropyl propanedioate, malonic acid, isobutyl-, dimethyl ester, acmc-1ailj, dimethyl2-isobutylmalonate
170218
dimethyl 2-(2-methylpropyl)propanedioate
COC(=O)C(CC(C)C)C(=O)OC
39520-24-6
C9H16O4
188.22
MFCD00015640
GAZFDPSEEIVCEX-UHFFFAOYSA-N
dimethyl isobutylmalonate, dimethyl 2-isobutylmalonate, dimethylisobutylmalonate, propanedioic acid, 2-2-methylpropyl-, 1,3-dimethyl ester, propanedioic acid, 2-methylpropyl-, dimethyl ester, 1,3-dimethyl 2-2-methylpropyl propanedioate, malonic acid, isobutyl-, dimethyl ester, acmc-1ailj, dimethyl2-isobutylmalonate
170218
dimethyl 2-(2-methylpropyl)propanedioate
COC(=O)C(CC(C)C)C(=O)OC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 211.0°C (763.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4230 to 1.4250
Quantity 5 g
Formula Weight 188.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl isobutylmalonate, 98%
Show More Show Less
EINECSNumber 254-490-8
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit