Alfa Aesar™ Dimethyl isopropenyl phosphate, 95%

Manufacturer: Alfa Aesar™ H2735322

Quantity 100g

 View more versions of this product

Catalog No. AAH2735322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4185-82-4
    Molecular Formula C5H11O4P
    Formula Weight 166.11
    MDL Number MFCD09953485
    Synonym Dimethyl 1-methylethenyl phosphate
    Chemical Name or Material Dimethyl isopropenyl phosphate

    Specifications

    Formula Weight 166.11
    Molecular Formula C5H11O4P
    Boiling Point 120°C (7.5mmHg)
    MDL Number MFCD09953485
    CAS 4185-82-4
    Quantity 100g
    Synonym Dimethyl 1-methylethenyl phosphate
    Chemical Name or Material Dimethyl isopropenyl phosphate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit