Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl itaconate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
617-52-7
C7H10O4
158.15
ZWWQRMFIZFPUAA-UHFFFAOYSA-N
dimethyl itaconate,dimethyl 2-methylenesuccinate,dimethylitaconate,itaconic acid dimethyl ester,itaconic acid, dimethyl ester,butanedioic acid, methylene-, dimethyl ester,1,4-dimethyl 2-methylidenebutanedioate,dimethyl methylenesuccinate,unii-11jib0yi93,succinic acid, methylene-, dimethyl ester
69240
dimethyl 2-methylidenebutanedioate
COC(=O)CC(=C)C(=O)OC
617-52-7
C7H10O4
158.15
ZWWQRMFIZFPUAA-UHFFFAOYSA-N
dimethyl itaconate,dimethyl 2-methylenesuccinate,dimethylitaconate,itaconic acid dimethyl ester,itaconic acid, dimethyl ester,butanedioic acid, methylene-, dimethyl ester,1,4-dimethyl 2-methylidenebutanedioate,dimethyl methylenesuccinate,unii-11jib0yi93,succinic acid, methylene-, dimethyl ester
69240
dimethyl 2-methylidenebutanedioate
COC(=O)CC(=C)C(=O)OC
Specific Gravity 1.24
Boiling Point 208.0°C
Quantity 5g
Packaging Glass bottle
Density 1.2400g/mL
Melting Point 34.0°C to 39.0°C
Color White
Infrared Spectrum Authentic
Flash Point 100°C
Assay Percent Range 97%
Linear Formula CH3O2CCH2C(=CH2)CO2CH3
Beilstein 02,762
Solubility Information Solubility in water: slightly soluble
Formula Weight 158.15
Physical Form Crystalline Low Melting Mass or Liquid
Percent Purity 97%
Chemical Name or Material Dimethyl itaconate
Show More Show Less