Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Dimethyl-β-methylsuccinimide, 99+%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
61748-86-5
C7H11NO2
141.17
MFCD00005497
SUPYQTKKYLUOBW-UHFFFAOYSA-N
2,2,3-trimethylsuccinimide,alpha,alpha-dimethyl-beta-methylsuccinimide,a,a-dimethyl-b-methylsuccinimide,supyqtkkyluobw-uhfffaoysa,3,3,4-trimethylazolidine-2,5-dione,2,5-pyrrolidinedione, 3,3,4-trimethyl,3,3,4-trimethyl-2,5-pyrrolidinedione #,.alpha.,.alpha.-dimethyl-.beta.-methylsuccinimide
550809
3,3,4-trimethylpyrrolidine-2,5-dione
CC1C(=O)NC(=O)C1(C)C
61748-86-5
C7H11NO2
141.17
MFCD00005497
SUPYQTKKYLUOBW-UHFFFAOYSA-N
2,2,3-trimethylsuccinimide,alpha,alpha-dimethyl-beta-methylsuccinimide,a,a-dimethyl-b-methylsuccinimide,supyqtkkyluobw-uhfffaoysa,3,3,4-trimethylazolidine-2,5-dione,2,5-pyrrolidinedione, 3,3,4-trimethyl,3,3,4-trimethyl-2,5-pyrrolidinedione #,.alpha.,.alpha.-dimethyl-.beta.-methylsuccinimide
550809
3,3,4-trimethylpyrrolidine-2,5-dione
CC1C(=O)NC(=O)C1(C)C
Packaging Glass Bottle
Quantity 250mg
Color White
Melting Point 117.0°C to 122.0°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Beilstein 21, 394
Formula Weight 141.17
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material α, α-Dimethyl-β-methylsuccinimide
Show More Show Less

EINECSNumber : 262-956-7

TSCA : TSCA