Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl oxazole-4,5-dicarboxylate, 99%, Thermo Scientific™

Catalog No. AAH3000903
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The teroxazole has been synthesized from dimethyl oxazole-4,5-dicarboxylate and lithiobenzyl isocyanide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72030-81-0
Molecular Formula C7H7NO5
Molecular Weight (g/mol) 185.135
MDL Number MFCD05863258
InChI Key LYRDBSFJJDATFO-UHFFFAOYSA-N
Synonym dimethyl oxazole-4,5-dicarboxylate, 4,5-dimethyl 1,3-oxazole-4,5-dicarboxylate, methyl 5-methoxycarbonyl-1,3-oxazole-4-carboxylate
PubChem CID 819934
IUPAC Name dimethyl 1,3-oxazole-4,5-dicarboxylate
SMILES COC(=O)C1=C(OC=N1)C(=O)OC

Specifications

Melting Point 74°C to 76°C
Boiling Point 150°C (12 mmHg)
Quantity 1 g
Formula Weight 185.14
Percent Purity 99%
Chemical Name or Material Dimethyl oxazole-4,5-dicarboxylate
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.