Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl oxazole-4,5-dicarboxylate, 99%, Thermo Scientific™
Click to view available options
Quantity:
1g
10g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
The teroxazole has been synthesized from dimethyl oxazole-4,5-dicarboxylate and lithiobenzyl isocyanide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
The teroxazole has been synthesized from dimethyl oxazole-4,5-dicarboxylate and lithiobenzyl isocyanide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 74°C to 76°C |
Boiling Point | 150°C (12mmHg) |
Quantity | 10g |
Formula Weight | 185.14 |
Percent Purity | 99% |
Chemical Name or Material | Dimethyl oxazole-4,5-dicarboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures