Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Phthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal drum
131-11-3
C10H10O4
194.19
MFCD00008425
NIQCNGHVCWTJSM-UHFFFAOYSA-N
dimethyl phthalate,dimethylphthalate,solvanom,solvarone,avolin,fermine,phthalic acid dimethyl ester,mipax,palatinol m,unimoll dm
8554
CHEBI:4609
dimethyl benzene-1,2-dicarboxylate
COC(=O)C1=CC=CC=C1C(=O)OC
131-11-3
C10H10O4
194.19
MFCD00008425
NIQCNGHVCWTJSM-UHFFFAOYSA-N
dimethyl phthalate,dimethylphthalate,solvanom,solvarone,avolin,fermine,phthalic acid dimethyl ester,mipax,palatinol m,unimoll dm
8554
CHEBI:4609
dimethyl benzene-1,2-dicarboxylate
COC(=O)C1=CC=CC=C1C(=O)OC
Boiling Point 282°C
Flash Point 150°C
Specific Gravity 1.19
Viscosity 17 mPa.s (20°C)
Density 1.1900g/mL
Refractive Index 1.5140 to 1.516
Infrared Spectrum Authentic
Quantity 250mL
Packaging Glass bottle
Color Undesignated
Assay Percent Range 99%
Beilstein 09, 797
Merck Index 15, 3279
Solubility Information Solubility in water: negligible. Other solubilities: miscible with organic solvents,insoluble in paraffinic hydrocarbons,slightly soluble in mineral oil
Formula Weight 194.19
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Dimethyl phthalate
Show More Show Less