Alfa Aesar™ Dimethyl pimelate, 99%

Manufacturer: Alfa Aesar™ B2202622

Quantity 100g

 View more versions of this product

Catalog No. AAB2202622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1732-08-7
    Molecular Formula C9H16O4
    Linear Formula CH3O2C(CH2)5CO2CH3
    Formula Weight 188.23
    MDL Number MFCD00008470
    Beilstein 1777309
    Synonym Pimelic acid dimethyl ester; Dimethyl heptanedioate
    Chemical Name or Material Dimethyl pimelate

    Specifications

    Formula Weight 188.23
    TSCA TSCA
    Linear Formula CH3O2C(CH2)5CO2CH3
    MDL Number MFCD00008470
    Flash Point >110°C (230°F)
    Odor Mild
    Boiling Point 121° to 122°C (11mmHg)
    Quantity 100g
    Molecular Formula C9H16O4
    CAS 1732-08-7
    Beilstein 1777309
    Synonym Pimelic acid dimethyl ester; Dimethyl heptanedioate
    Chemical Name or Material Dimethyl pimelate
    Assay 99%

    Description

    Applications
    Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

    Solubility
    Soluble in ethanol, benzene, ether. Slightly soluble in water.

    Notes
    Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit