Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl pyridine-2,3-dicarboxylate, 95%

Click to view available options
Quantity:
5g
25g
100g
605-38-9
C9H9NO4
195.174
MFCD00186549
YLGIBCYHQZTFQL-UHFFFAOYSA-N
2,3-pyridinedicarboxylic acid dimethyl ester,dimethyl quinolinate,dimethyl 2,3-pyridinedicarboxylate,2,3-bis methoxycarbonyl pyridine,2,3-pyridinedicarboxylic acid, dimethyl ester,methylquinolinic acid, 2,3-dimethyl ester,2,3-dimethyl pyridine-2,3-dicarboxylate,pyridine-2,3-dicarboxylic acid dimethyl ester,mqdme,2,3-dimethyl methylquinolinate
69058
dimethyl pyridine-2,3-dicarboxylate
COC(=O)C1=C(N=CC=C1)C(=O)OC
605-38-9
C9H9NO4
195.174
MFCD00186549
YLGIBCYHQZTFQL-UHFFFAOYSA-N
2,3-pyridinedicarboxylic acid dimethyl ester,dimethyl quinolinate,dimethyl 2,3-pyridinedicarboxylate,2,3-bis methoxycarbonyl pyridine,2,3-pyridinedicarboxylic acid, dimethyl ester,methylquinolinic acid, 2,3-dimethyl ester,2,3-dimethyl pyridine-2,3-dicarboxylate,pyridine-2,3-dicarboxylic acid dimethyl ester,mqdme,2,3-dimethyl methylquinolinate
69058
dimethyl pyridine-2,3-dicarboxylate
COC(=O)C1=C(N=CC=C1)C(=O)OC
Melting Point 53°C to 58°C
Quantity 100g
Beilstein 159846
Formula Weight 195.17
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Dimethyl pyridine-2,3-dicarboxylate