Alfa Aesar™ Dimethyl selenide

Manufacturer: Alfa Aesar™ 6811614

Quantity 25g

 View more versions of this product

Catalog No. AA6811614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 593-79-3
    Molecular Formula C2H6Se
    Linear Formula (CH3)2Se
    Formula Weight 109.03
    MDL Number MFCD00014848
    UN Number UN3282
    Beilstein 1696848
    Synonym Methyl selenide
    Chemical Name or Material Dimethyl selenide

    Specifications

    Formula Weight 109.03
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Methyl selenide
    RTECS Number VS6880000
    Molecular Formula C2H6Se
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Odor Unpleasant
    Sensitivity Air & Light Sensitive
    Boiling Point 56° to 58°C
    Linear Formula (CH3)2Se
    Beilstein 1696848
    Health Hazard 1 Danger
    Recommended Storage 2°C to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, TOXIC, LIQUID, N.O.S.
    Quantity 25g
    Physical Form Liquid
    MDL Number MFCD00014848
    CAS 593-79-3
    Density 1.41g/mL
    Color Colorless
    UN Number UN3282
    Chemical Name or Material Dimethyl selenide

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    2°C to 8°C

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit