Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5L
Packaging:
Plastic bottle
77-78-1
C2H6O4S
126.13
MFCD00008416
VAYGXNSJCAHWJZ-UHFFFAOYSA-N
dimethyl sulphate,sulfuric acid, dimethyl ester,dimethyl monosulfate,dimethylsulfate,dimethylsulfat,sulfate dimethylique,sulfuric acid dimethyl ester,dimethylsulfaat,dimetilsolfato,dms methyl sulfate
6497
CHEBI:59050
dimethyl sulfate
COS(=O)(=O)OC
77-78-1
C2H6O4S
126.13
MFCD00008416
VAYGXNSJCAHWJZ-UHFFFAOYSA-N
dimethyl sulphate,sulfuric acid, dimethyl ester,dimethyl monosulfate,dimethylsulfate,dimethylsulfat,sulfate dimethylique,sulfuric acid dimethyl ester,dimethylsulfaat,dimetilsolfato,dms methyl sulfate
6497
CHEBI:59050
dimethyl sulfate
COS(=O)(=O)OC
Alkylation reagent
Viscosity 1.81 mPa.s (20°C)
Specific Gravity 1.33
Density 1.3300g/mL
Refractive Index 1.3860 to 1.3880
Packaging Plastic bottle
Flash Point 83°C
Color Colorless
Melting Point -32.0°C
Boiling Point 188.0°C
Quantity 5L
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula (CH3)2SO4
Beilstein 01,283
Fieser 01,293; 04,188; 13,239
Merck Index 15,328
Solubility Information Solubility in water: 2.8g/100mL (18°C). Other solubilities: hydrolysis rapidly at 18°C or above in water,soluble in ehter,dioxane,acetone and aromatic,hydrocarbons,sparingly soluble in carbon disulfide and,aliphatic hydrocarbons
Formula Weight 126.13
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Dimethyl sulfate
Show More Show Less

GHS H StatementMay cause cancer.Suspected of causing genetic defects.Fatal if inhaled.Toxic if swallowed.Causes severe skin burns and eye damage.May cause an allergic skin reaction.May cause respiratory i

GHS P StatementIF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Immediately call a POISON CENTER or doctor/physician.Wear protective gloves/protective clothing/eye protection/face protection.IF SWALLOWED: Rinse mouth.Do NOT induce vomiting.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.

;GHS P StatementIF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Immediately call a POISON CENTER or doctor/physician.Wear protective gloves/protective clothing/eye protection/face protection.IF SWALL