Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
67-68-5
67-68-5
Packaging Amber Glass Bottle
Acidity 0.0004
Quantity 500 mL
CAS Max % 1
Percent Purity 0.995
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.