Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, ACS, 99.9% min, Alfa Aesar™

Click to view available options
Quantity:
500mL
1L
4L
4 x 1L
4 x 4L
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
Assay Percent Range 99.9%
Beilstein 506008
Solubility Information Miscible with water,diethyl ether,ethanol,acetone,ether,carbon tetrachloride and ethyl acetate.
Formula Weight 78.13
Physical Form Clear,colorless Liquid
Grade ACS
Density 1.101 g/mL
Refractive Index 1.479
Flash Point 87°C (189°F)
Boiling Point 189°C
Color Colorless
Melting Point 18.4°C
Quantity 500mL
Sensitivity Hygroscopic
Chemical Name or Material Dimethyl sulfoxide, For analysis ACS
Show More Show Less

P210-P370+P378q-P501c

H227

EINECSNumber : 200-664-3

RTECSNumber : PV6210000

TSCA : Yes

Recommended Storage : Ambient temperatures