Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, ACS Grade, OmniPur™ Calbiochem™

Click to view available options
Quantity:
500mL
67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C
67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C
Percent Purity ≥99.9%
Grade ACS
Quantity 500mL