Learn More
Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
Dimethyl sulfoxide is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. DMSO is an effective pain relieving agent. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids. Used to enhance dermal absorption of many chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 18.4°C |
Color | Colorless |
Assay Percent Range | >99.8% |
Beilstein | 506008 |
Solubility Information | Miscible with water,diethyl ether,ethanol,acetone,ether,carbon tetrachloride and ethyl acetate. |
IUPAC Name | methylsulfinylmethane |
Formula Weight | 78.13 |
Density | 1.101 g/cm3 at 20°C |
Boiling Point | 189°C |
Quantity | 100 mL |
Show More |
Safety and Handling
Hazard Category | H227 |
Precautionary Statement | P210-P370+P378q-P501c |
EINECSNumber | 200-664-3 |
RTECSNumber | PV6210000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.