Dimethyl sulfoxide, anhydrous, >99.8%, Alfa Aesar™

Manufacturer: Alfa Aesar 43998K2

Quantity 1L

 View more versions of this product

Catalog No. AA43998K2

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 67-68-5
    Physical Form Liquid
    Assay Percent Range >99.8%
    Molecular Formula (CH3)2SO
    Formula Weight 78.13
    MDL Number MFCD00002089
    Chemical Name or Material Dimethyl sulfoxide

    Specifications

    Boiling Point 189°C
    Physical Form Liquid
    Color Clear colorless
    Melting Point 18.4°C
    Formula Weight 78.13
    Quantity 1L
    Flash Point 87°C (188.6°F)
    EINECS Number 200-664-3
    Odor Mild
    Molecular Formula (CH3)2SO
    MDL Number MFCD00002089
    Ignition Point 270°C (518°F)
    Vapor Pressure 2mm Hg at 20°C
    Assay Percent Range >99.8%
    CAS 67-68-5
    Health Hazard 1 H227: Combustible liquid.
    Chemical Name or Material Dimethyl sulfoxide

    H227: Combustible liquid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit