Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Poly Pail

Chemical Identifiers

67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CHEBI:28262

High quality chemicals for laboratory and specialized industrial use

SMILES CS(=O)C
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 1.10kg
Flash Point 89.00°C
Quantity 4L
Chemical Name or Material Dimethyl Sulfoxide
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements