Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Poly Pail
67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C
67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C

High quality chemicals for laboratory and specialized industrial use

Formula Weight 78.13
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Flash Point 89.00°C
Quantity 500mL
Chemical Name or Material Dimethyl Sulfoxide