Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl sulfoxide, Bioreagent

Click to view available options
Quantity:
50mL
100mL
250mL
500mL
1L
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
Applications
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.

Solubility
Soluble in water, methanol, acetone, ether, benzene, chloroform.

Notes
Submicron filtered. Store at Room Temperature (15-30°C), desiccate. Hygroscopic.
Beilstein 506008
Solubility Information Soluble in water,methanol,acetone,ether,benzene,chloroform.
Formula Weight 78.13
Physical Form Liquid
Density 1.101 g/mL
Odor Mild
Refractive Index 1.479
Flash Point 87°C (189°F)
Boiling Point 189°C
Color Colorless
Melting Point 18.4°C
Quantity 50mL
Sensitivity Hygroscopic
Chemical Name or Material Dimethyl sulfoxide, Bioreagent
Show More Show Less

GHS H Statement
H227
Combustible liquid.

P210-P370+P378q-P501c

H227

EINECSNumber : 200-664-3

RTECSNumber : PV6210000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon