Learn More
Dimethyl sulfoxide-d{6}, 99.5% (Isotopic), Thermo Scientific Chemicals
Description
DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.
Solubility
Miscible with water and with almost all organic solvents such as alcohols, esters, ketones, chlorinated solvents and aromatic hydrocarbons.
Notes
NMR solvent. Dimethyl Sulfoxide (DMSO) has a melting point of 20°C, freezing close to room temperature. Sometimes when it is delivered it will be in a solid state. To return the material to a liquid state, thaw it in a warm water bath. Care must be taken to prevent water contamination. It is stable, combustible and moisture sensitive. Incompatible with acid chlorides, strong acids, phosphorus halides, strong oxidizing agents and strong reducing agents.
Specifications
Quantity | 10 g |
Refractive Index | 1.476 |
Sensitivity | Hygroscopic |
Flash Point | 88°C (190°F) |
Density | 1.18 g/mL |
Melting Point | 20°C |
Boiling Point | 190°C |
Assay Percent Range | 99.5% |
Solubility Information | Miscible with water and with almost all organic solvents such as alcohols,esters,ketones,chlorinated solvents and aromatic hydrocarbons. |
Formula Weight | 84.17 |
Show More |
Safety and Handling
Hazard Category | H227 |
Precautionary Statement | P210-P280-P370+P378q-P501c |
EINECSNumber | 218-617-0 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.