Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide-d6, 99.9 Atom% D, Spectrum™ Chemical
SDP

D2493, 2206-27-1, CD3SOCD3

$84.00 - $563.00

Specifications

CAS 2206-27-1
CAS Max % 100%
Grade Reagent
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-602-247
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
D24935GM
5 g Amber Glass Bottle
Each for $84.00
Add to Cart
 
18-602-252
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
D2493135
10 x 0.75 mL Ampule
Each for $110.00
Add to Cart
 
18-602-248
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
D249310GM
10 g Amber Glass Bottle
Each for $133.00
Add to Cart
 
18-602-246
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
D2493136
10 x 1 g Ampule
Each for $150.00
Add to Cart
 
18-602-249
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
D249325GM
25 g Amber Glass Bottle
Each for $307.00
Add to Cart
 
18-602-250
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
D249350GM
50 g Amber Glass Bottle
Each for $563.00
Add to Cart
 
Description

Description

Spectrum™ Chemical Dimethyl Sulfoxide-d6, 99.9 Atom Percent D - Spectrum offers a broad range of high quality deuterated products for all your routine synthesis and structural analysis needs and is the preferred brand for deuterated NMR solvents worldwide. We offer Isotopic Enrichment ranges from 97 atom % D for routine use to 100 atom % D for the most demanding applications.
Specifications

Specifications

2206-27-1
100%
Reagent
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit