Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
6 x 1 L
200 L
67-68-5
C2H6OS
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methanesulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methanesulfinylmethane
CS(C)=O

HP582, 67-68-5, C2H6OS

Spectrum™ Dimethyl Sulfoxide, Exceeds A.C.S. Specifications, HPLC Grade The GR is an EMD trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.
Packaging Amber Glass Bottle
CAS Max % 1
Percent Purity 99.9%
Residue after Evaporation 5 ppm
Water 0.0005
Quantity 4 L