Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, HPLC Grade, 99.9%, Alfa Aesar™

Click to view available options
Quantity:
250mL
1L
4 x 1L
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O

Sensitivity: Hygroscopic

Appearance Colorless liquid
Assay Percent Range >99.9%
Beilstein 506008
Solubility Information Miscible with water,diethyl ether,ethanol,acetone,ether,carbon tetrachloride and ethyl acetate.
Formula Weight 78.13
Physical Form Liquid
Grade HPLC
Density 1.101 g/cm3 at 20°C
Ignition Point 270°C (518°F)
Odor Mild
Refractive Index 1.479
Vapor Pressure 2.5 hPa (2mm Hg) at 20°C
Flash Point 87°C (189°F)
Boiling Point 189°C
Color Colorless
Melting Point 18.4°C
Quantity 250mL
Sensitivity Hygroscopic
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less

P210-P370+P378q-P501c

H227

EINECSNumber : 200-664-3

RTECSNumber : PV6210000

TSCA : Yes

Recommended Storage : Ambient temperatures