Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, PHOTREX™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CHEBI:28262

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry
CAS 67-68-5
Molecular Formula C2H6OS
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(=O)C
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.129
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Flash Point 89.00°C
Quantity 4L
Chemical Name or Material Dimethyl Sulfoxide
Grade PHOTREX™ Reagent
Show More Show Less