Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, SpectroSolv (TM), 99.9%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
67-68-5
67-68-5
CAS Max % 1
Appearance Clear, Colorless
Percent Purity 0.999
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Titratable Acid 1 μeq/g
Water 0.0005
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.