Alfa Aesar™ Dimethyl terephthalate-2,3,5,6-d4, 98 atom % D

Manufacturer: Alfa Aesar™ H51104MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51104MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74079-01-9
    Molecular Formula C10D4H6O4
    Formula Weight 198.21
    MDL Number MFCD00182544
    Synonym 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester; Dimethyl terephthalate-d4(ring-d4)
    Chemical Name or Material Dimethyl terephthalate-2,3,5,6-d4

    Specifications

    Formula Weight 198.21
    Molecular Formula C10D4H6O4
    MDL Number MFCD00182544
    CAS 74079-01-9
    Quantity 250mg
    Synonym 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester; Dimethyl terephthalate-d4(ring-d4)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Dimethyl terephthalate-2,3,5,6-d4
    Assay 95%, 98 atom % D

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit