Alfa Aesar™ Dimethyl tert-butylmalonate, 95%

Manufacturer: Alfa Aesar™ H2653206

Quantity 5g

 View more versions of this product

Catalog No. AAH2653206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39520-25-7
    Molecular Formula C9H16O4
    Formula Weight 188.22
    MDL Number MFCD08436110
    Synonym tert-Butylmalonic acid dimethyl ester
    Chemical Name or Material Dimethyl tert-butylmalonate

    Specifications

    Formula Weight 188.22
    MDL Number MFCD08436110
    Molecular Formula C9H16O4
    CAS 39520-25-7
    Quantity 5g
    Synonym tert-Butylmalonic acid dimethyl ester
    Chemical Name or Material Dimethyl tert-butylmalonate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit