Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl (trityloxyimino)malonate, 98%

Click to view available options
Quantity:
250mg
1g
5g
82874-83-7
(Trityloxyimino)malonic acid dimethyl ester; Dimethyl 2-[(triphenylmethoxy)imino]malonate
82874-83-7
(Trityloxyimino)malonic acid dimethyl ester; Dimethyl 2-[(triphenylmethoxy)imino]malonate
Quantity 5g
Formula Weight 403.43
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Dimethyl (trityloxyimino)malonate

TSCA : No

Recommended Storage : Ambient temperatures